4,6-Dimethoxy-5-methylpyrimidine 95%

Lieferant: Apollo Scientific

OR951178-1G OR951178-250MG
APOSOR951178-1GEA 149 EUR
APOSOR951178-1G APOSOR951178-250MG
4,6-Dimethoxy-5-methylpyrimidine 95%
4,6-Dimethoxy-5-methylpyrimidine
Formel: C₇H₁₀N₂O₂
Molecular Weight: 154.166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00196466
CAS-Nummer: 13566-63-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR