(S)-[(1-Phenylethyl)amino]acetic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-2-((1-Phenylethyl)amino)acetic acid

OR951255-25G OR951255-5G
APOSOR951255-25GEA 150 EUR
APOSOR951255-25G APOSOR951255-5G
(S)-[(1-Phenylethyl)amino]acetic acid 98%
(S)-[(1-Phenylethyl)amino]acetic acid
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11656778
CAS-Nummer: 78397-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR