3-Methyl-1-(pyridin-2-yl)butan-1-amine 95%
Lieferant: Apollo Scientific
OR951278-1G
OR951278-250MG
APOSOR951278-1GEA
364
EUR
APOSOR951278-1G
APOSOR951278-250MG
3-Methyl-1-(pyridin-2-yl)butan-1-amine 95%
3-Methyl-1-(pyridin-2-yl)butan-1-amine
Formel:
C₁₀H₁₆N₂ Molecular Weight: 164,247 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12153903 CAS-Nummer: 825647-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...