2-Cyano-3-phenyl-acrylic acid methyl ester 95%

Lieferant: Apollo Scientific

Synonyme: Methyl (E)-2-Cyano-3-phenylacrylate

OR951313-1G OR951313-5G
APOSOR951313-1GEA 92.8 EUR
APOSOR951313-1G APOSOR951313-5G
2-Cyano-3-phenyl-acrylic acid methyl ester 95%
2-Cyano-3-phenyl-acrylic acid methyl ester
Formel: C₁₁H₉NO₂
Molecular Weight: 187,195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00025864
CAS-Nummer: 14533-86-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR