D-N-Benzylserine methyl ester 96%
Lieferant: Apollo Scientific
Synonyme:
N-Benzyl-D-Serine-OMe Ester, Methyl (2R)-2-(benzylamino)-3-hydroxypropanoate, D-N-Benzylserine Methyl Ester
OR951335-1G
OR951335-5G
APOSOR951335-1GEA
159
EUR
APOSOR951335-1G
APOSOR951335-5G
D-N-Benzylserine methyl ester 96%
D-N-Benzylserine methyl ester
Formel:
C₁₁H₁₅NO₃ Molecular Weight: 209,242 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD12198843 CAS-Nummer: 131110-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...