Pseudopelletierine oxime 98%

Lieferant: Apollo Scientific

Synonyme: 9-methyl-9-azabicyclo[3.3.1]nonan-3-one oxime

OR951413-100G OR951413-25G
APOSOR951413-100GEA 231 EUR
APOSOR951413-100G APOSOR951413-25G
Pseudopelletierine oxime 98%
Pseudopelletierine oxime
Formel: C₉H₁₆N₂O
Molecular Weight: 168.236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03755545
CAS-Nummer: 6164-67-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR