4-(1,3,4-Oxadiazol-2-yl)benzoic acid 95%
Lieferant: Apollo Scientific
OR951426-10G
OR951426-1G
OR951426-5G
APOSOR951426-10GEA
502
EUR
APOSOR951426-10G
APOSOR951426-1G
APOSOR951426-5G
4-(1,3,4-Oxadiazol-2-yl)benzoic acid 95%
4-(1,3,4-Oxadiazol-2-yl)benzoic acid
Formel:
C₉H₆N₂O₃ Molecular Weight: 190,156 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07352166 CAS-Nummer: 59663-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...