1-[4-(1H-Imidazol-1-yl)phenyl]ethanamine 95%
Lieferant: Apollo Scientific
OR951450-1G
OR951450-250MG
OR951450-5G
APOSOR951450-1GEA
290
EUR
APOSOR951450-1G
APOSOR951450-250MG
APOSOR951450-5G
1-[4-(1H-Imidazol-1-yl)phenyl]ethanamine 95%
1-[4-(1H-Imidazol-1-yl)phenyl]ethanamine
Formel:
C₁₁H₁₃N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04970993 CAS-Nummer: 704877-65-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...