5-Chloro-2-furoic acid 98%
Lieferant: Apollo Scientific
Synonyme:
5-Chlorofuran-2-carboxylic acid
OR951451-5G
APOSOR951451-5GEA
58
EUR
APOSOR951451-5G
5-Chloro-2-furoic acid 98%
5-Chloro-2-furoic acid
Formel:
C₅H₃ClO₃ Molecular Weight: 146,529 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00093272 CAS-Nummer: 618-30-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...