Methyl 4-pyridinylacetate

Lieferant: Apollo Scientific

Synonyme: Methyl 2-(pyridin-4-yl)acetate

OR951455-25G
APOSOR951455-25GEA 325 EUR
APOSOR951455-25G
Methyl 4-pyridinylacetate
Methyl 4-pyridinylacetate
Formel: C₈H₉NO₂
Molecular Weight: 151,163 g/mol
Siedepunkt: 150 °C
Schmelzpunkt: 8…8,5 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00972014
CAS-Nummer: 29800-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR