Ethyl alfa-amino butyrate hydrochloride 98%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-Aminobutyrate Hydrochloride

OR951485-5G
APOSOR951485-5GEA 105 EUR
APOSOR951485-5G
Ethyl alfa-amino butyrate hydrochloride 98%
Ethyl alfa-amino butyrate hydrochloride
Formel: C₆H₁₃NO₂·ClH
Molecular Weight: 167,634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08689937
CAS-Nummer: 55410-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR