Ethyl 5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate 95%
Lieferant: Apollo Scientific
OR951571-1G OR951571-250MG
APOSOR951571-1GEA 393 EUR
APOSOR951571-1G APOSOR951571-250MG
Ethyl 5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate 95%
Ethyl 5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate
Formel: C₉H₁₃N₃O₂
Molecular Weight: 195.218 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08059257
CAS-Nummer: 91476-82-3