1H-Pyrrolo[2,3-c]pyridine-5-carbaldehyde 95%

Lieferant: Apollo Scientific

OR951602-1G OR951602-250MG
APOSOR951602-1GEA 243 EUR
APOSOR951602-1G APOSOR951602-250MG
1H-Pyrrolo[2,3-c]pyridine-5-carbaldehyde 95%
1H-Pyrrolo[2,3-c]pyridine-5-carbaldehyde
Formel: C₈H₆N₂O
Molecular Weight: 146,146 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10697530
CAS-Nummer: 130473-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR