3-Methyl-2-oxo-butanoic acid 95%
Lieferant: Apollo Scientific
Synonyme:
3-Methyl-2-oxobutanoic acid
OR951671-1G
APOSOR951671-1GEA
72.5
EUR
APOSOR951671-1G
3-Methyl-2-oxo-butanoic acid 95%
3-Methyl-2-oxo-butanoic acid
Formel:
C₅H₈O₃ Molecular Weight: 116,115 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040427 CAS-Nummer: 759-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...