4-Hydroxyquinoline-3-carbonitrile 95%
Lieferant: Apollo Scientific
OR951730-1G
OR951730-250MG
APOSOR951730-1GEA
864
EUR
APOSOR951730-1G
APOSOR951730-250MG
4-Hydroxyquinoline-3-carbonitrile 95%
4-Hydroxyquinoline-3-carbonitrile
Formel:
C₁₀H₆N₂O Molecular Weight: 170.167 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07787027 CAS-Nummer: 2305-70-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...