(Phenylcyclopropyl)methylamine hydrochloride 98%

Lieferant: Apollo Scientific

Synonyme: (1-Phenylcyclopropyl)methylamine Hydrochloride

OR951733-1G OR951733-5G
APOSOR951733-1GEA 161 EUR
APOSOR951733-1G APOSOR951733-5G
(Phenylcyclopropyl)methylamine hydrochloride 98%
(Phenylcyclopropyl)methylamine hydrochloride
Formel: C₁₀H₁₃N·ClH
Molecular Weight: 183,678 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00034947
CAS-Nummer: 935-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR