1-Methyl-5-phenyl-1H-imidazole-2-thiol 95%

Lieferant: Apollo Scientific

OR951748-1G OR951748-250MG
APOSOR951748-1GEA 538 EUR
APOSOR951748-1G APOSOR951748-250MG
1-Methyl-5-phenyl-1H-imidazole-2-thiol 95%
1-Methyl-5-phenyl-1H-imidazole-2-thiol
Formel: C₁₀H₁₀N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD01429053
CAS-Nummer: 25433-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR