N'-(3-Nitropyridin-4-yl)acetohydrazide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(N'-Acetylhydrazino)-3-nitropyridine, Acetic acid N'-(3-nitropyridin-4-yl)hydrazide
OR9519-1G
OR9519-5G
APOSOR9519-1GEA
133
EUR
APOSOR9519-1G
APOSOR9519-5G
N'-(3-Nitropyridin-4-yl)acetohydrazide
N'-(3-Nitropyridin-4-yl)acetohydrazide
Formel:
C₇H₈N₄O₃ Molecular Weight: 196,163 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00956008 CAS-Nummer: 31481-85-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...