5-Phenyl-1H-pyrimidine-2,4-dione 98%
Lieferant: Apollo Scientific
OR951934-1G
OR951934-250MG
APOSOR951934-1GEA
493
EUR
APOSOR951934-1G
APOSOR951934-250MG
5-Phenyl-1H-pyrimidine-2,4-dione 98%
5-Phenyl-1H-pyrimidine-2,4-dione
Formel:
C₁₀H₈N₂O₂ Molecular Weight: 188,183 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06801865 CAS-Nummer: 15761-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...