2,3-Dihydro-1,4-benzodioxin-6-carboxylic acid methyl ester 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 1,4-Benzodioxan-6-carboxylate

OR951985-25G OR951985-5G
APOSOR951985-25GEA 455 EUR
APOSOR951985-25G APOSOR951985-5G
2,3-Dihydro-1,4-benzodioxin-6-carboxylic acid methyl ester 98%
2,3-Dihydro-1,4-benzodioxin-6-carboxylic acid methyl ester
Formel: C₁₀H₁₀O₄
Molecular Weight: 194.184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01544907
CAS-Nummer: 20197-75-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR