Methyl 3-methyl-2-oxobutanoate 95%

Lieferant: Apollo Scientific

OR952085-1G OR952085-250MG
APOSOR952085-1GEA 574 EUR
APOSOR952085-1G APOSOR952085-250MG
Methyl 3-methyl-2-oxobutanoate 95%
Methyl 3-methyl-2-oxobutanoate
Formel: C₆H₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12198846
CAS-Nummer: 3952-67-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR