Benzo[b]thiophene-3(2H)-one 1,1-dioxide 95%
Lieferant: Apollo Scientific
OR952294-1G
OR952294-25G
OR952294-5G
APOSOR952294-1GEA
0
EUR
APOSOR952294-1G
APOSOR952294-25G
APOSOR952294-5G
Benzo[b]thiophene-3(2H)-one 1,1-dioxide 95%
Benzo[b]thiophene-3(2H)-one 1,1-dioxide
Formel:
C₈H₆O₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00159164 CAS-Nummer: 1127-35-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...