6-Cyclopropyl-2,3-dihydropyridazin-3-one 95%

Lieferant: Apollo Scientific

Synonyme: 6-Cyclopropylpyridazin-3(2H)-one

OR952331-1G OR952331-250MG
APOSOR952331-1GEA 101 EUR
APOSOR952331-1G APOSOR952331-250MG
6-Cyclopropyl-2,3-dihydropyridazin-3-one 95%
6-Cyclopropyl-2,3-dihydropyridazin-3-one
Formel: C₇H₈N₂O
Molecular Weight: 136,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16871334
CAS-Nummer: 1161737-33-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR