(2S,4R)-1-Boc-2-Cyano-4-hydroxypyrrolidine 98%

Lieferant: Apollo Scientific

Synonyme: (2S,4R)-tert-Butyl 2-cyano-4-hydroxypyrrolidine-1-carboxylate

OR952355-1G OR952355-25G OR952355-5G
APOSOR952355-1GEA 55.1 EUR
APOSOR952355-1G APOSOR952355-25G APOSOR952355-5G
(2S,4R)-1-Boc-2-Cyano-4-hydroxypyrrolidine 98%
(2S,4R)-1-Boc-2-Cyano-4-hydroxypyrrolidine
Formel: C₁₀H₁₆N₂O₃
Molecular Weight: 212,246 g/mol
Schmelzpunkt: 90…110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09863132
CAS-Nummer: 483366-12-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR