6-Methyl-7-oxabicyclo[4.1.0]heptan-2-one 95%

Lieferant: Apollo Scientific

OR952470-1G OR952470-250MG
APOSOR952470-1GEA 696 EUR
APOSOR952470-1G APOSOR952470-250MG
6-Methyl-7-oxabicyclo[4.1.0]heptan-2-one 95%
6-Methyl-7-oxabicyclo[4.1.0]heptan-2-one
Formel: C₇H₁₀O₂
Molecular Weight: 126.153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17926441
CAS-Nummer: 21889-89-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR