Methyl 4-methoxy-3-methylbenzoate 95%

Lieferant: Apollo Scientific

OR952508-1G OR952508-25G OR952508-5G
APOSOR952508-1GEA 125 EUR
APOSOR952508-1G APOSOR952508-25G APOSOR952508-5G
Methyl 4-methoxy-3-methylbenzoate 95%
Methyl 4-methoxy-3-methylbenzoate
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203809
CAS-Nummer: 70347-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR