7-Chloro-1H-indole-3-carboxylic acid 95%

Lieferant: Apollo Scientific

OR952586-1G OR952586-250MG
APOSOR952586-1GEA 364 EUR
APOSOR952586-1G APOSOR952586-250MG
7-Chloro-1H-indole-3-carboxylic acid 95%
7-Chloro-1H-indole-3-carboxylic acid
Formel: C₉H₆ClNO₂
Molecular Weight: 195,602 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07369968
CAS-Nummer: 86153-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR