7-Chloro-1H-quinazoline-2,4-dione 95%
Lieferant: Apollo Scientific
Synonyme:
7-Chloroquinazoline-2,4(1H,3H)-dione
OR952587-5G
APOSOR952587-5GEA
120
EUR
APOSOR952587-5G
7-Chloro-1H-quinazoline-2,4-dione 95%
7-Chloro-1H-quinazoline-2,4-dione
Formel:
C₈H₅ClN₂O₂ Molecular Weight: 196,591 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00563867 CAS-Nummer: 13165-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...