7-Hydroxy-1,2,3,4-tetrahydro-naphthalene-2-carboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: 7-Hydroxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid

OR952613-1G OR952613-250MG
APOSOR952613-1GEA 345 EUR
APOSOR952613-1G APOSOR952613-250MG
7-Hydroxy-1,2,3,4-tetrahydro-naphthalene-2-carboxylic acid 95%
7-Hydroxy-1,2,3,4-tetrahydro-naphthalene-2-carboxylic acid
Formel: C₁₁H₁₂O₃
Molecular Weight: 192.211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061210
CAS-Nummer: 31846-36-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR