Methyl 1-aminocyclohexanoate 95%

Lieferant: Apollo Scientific

OR952642-25G OR952642-5G
APOSOR952642-25GEA 192 EUR
APOSOR952642-25G APOSOR952642-5G
Methyl 1-aminocyclohexanoate 95%
Methyl 1-aminocyclohexanoate
Formel: C₈H₁₅NO₂
Molecular Weight: 157.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08686793
CAS-Nummer: 4507-57-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR