7-Methyl-3H-imidazo[4,5-b]pyridine 95%

Lieferant: Apollo Scientific

OR952650-1G OR952650-250MG
APOSOR952650-1GEA 770 EUR
APOSOR952650-1G APOSOR952650-250MG
7-Methyl-3H-imidazo[4,5-b]pyridine 95%
7-Methyl-3H-imidazo[4,5-b]pyridine
Formel: C₇H₇N₃
Molecular Weight: 133,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05723324
CAS-Nummer: 27582-20-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR