(1H-Pyrrol-2-yl)-acetic acid 95%

Lieferant: Apollo Scientific

OR952693-10G OR952693-1G OR952693-5G
APOSOR952693-10GEA 986 EUR
APOSOR952693-10G APOSOR952693-1G APOSOR952693-5G
(1H-Pyrrol-2-yl)-acetic acid 95%
(1H-Pyrrol-2-yl)-acetic acid
Formel: C₆H₇NO₂
Molecular Weight: 125,125 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01993666
CAS-Nummer: 79673-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR