3-Acetyl-6-chloroimidazo[1,2-b]pyridazine 95%
Lieferant: Apollo Scientific
OR952699-5G
APOSOR952699-5GEA
106
EUR
APOSOR952699-5G
3-Acetyl-6-chloroimidazo[1,2-b]pyridazine 95%
3-Acetyl-6-chloroimidazo[1,2-b]pyridazine
Formel:
C₈H₆ClN₃O Molecular Weight: 195,606 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11044766 CAS-Nummer: 90734-71-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...