1-Allyl-1H-benzimidazole-2-thiol 95%

Lieferant: Apollo Scientific

OR952740-100MG OR952740-1G OR952740-250MG
APOSOR952740-100MGEA 190 EUR
APOSOR952740-100MG APOSOR952740-1G APOSOR952740-250MG
1-Allyl-1H-benzimidazole-2-thiol 95%
1-Allyl-1H-benzimidazole-2-thiol
Formel: C₁₀H₁₀N₂S
Molecular Weight: 190,265 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06655204
CAS-Nummer: 87216-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR