(3-Cyanophenoxy)acetic acid 98%

Lieferant: Apollo Scientific

Synonyme: (3-Cyanophenoxy)acetic Acid

OR952746-1G OR952746-5G
APOSOR952746-1GEA 50.9 EUR
APOSOR952746-1G APOSOR952746-5G
(3-Cyanophenoxy)acetic acid 98%
(3-Cyanophenoxy)acetic acid
Formel: C₉H₇NO₃
Molecular Weight: 177.157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00516888
CAS-Nummer: 1879-58-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR