3-(Methylsulphonyl)benzyl alcohol 96%
Lieferant: Apollo Scientific
Synonyme:
(3-(Methylsulphonyl)phenyl)methanol
OR952819-1G
OR952819-250MG
APOSOR952819-1GEA
295
EUR
APOSOR952819-1G
APOSOR952819-250MG
3-(Methylsulphonyl)benzyl alcohol 96%
3-(Methylsulphonyl)benzyl alcohol
Formel:
C₈H₁₀O₃S Molecular Weight: 186,228 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15527275 CAS-Nummer: 220798-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...