Benzhydrylidene methylamine 95%

Lieferant: Apollo Scientific

Synonyme: N-(Diphenylmethylene)methanamine

OR952854-1G OR952854-5G
APOSOR952854-1GEA 154 EUR
APOSOR952854-1G APOSOR952854-5G
Benzhydrylidene methylamine 95%
Benzhydrylidene methylamine
Formel: C₁₄H₁₃N
Molecular Weight: 195,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02179401
CAS-Nummer: 13280-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR