2-Cyano-3-pyridinecarboxylic acid methyl ester 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-cyanonicotinate
OR952870-1G
APOSOR952870-1GEA
62.5
EUR
APOSOR952870-1G
2-Cyano-3-pyridinecarboxylic acid methyl ester 95%
2-Cyano-3-pyridinecarboxylic acid methyl ester
Formel:
C₈H₆N₂O₂ Molecular Weight: 162,145 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128863 CAS-Nummer: 75358-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...