4-Acetamido-3-ethoxynitrobenzene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(2-Ethoxy-4-nitrophenyl)acetamide
OR9529-1G
OR9529-5G
OR9529-25G
APOSOR9529-1GEA
21.9
EUR
APOSOR9529-1G
APOSOR9529-5G
APOSOR9529-25G
4-Acetamido-3-ethoxynitrobenzene
4-Acetamido-3-ethoxynitrobenzene
Formel:
C₁₀H₁₂N₂O₄ Molecular Weight: 224,213 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00994990 CAS-Nummer: 116496-76-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...