2-Amino-5-methoxycarbonylbenzoic acid 96%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-5-(methoxycarbonyl)benzoic acid

OR953029-1G OR953029-250MG
APOSOR953029-1GEA 604 EUR
APOSOR953029-1G APOSOR953029-250MG
2-Amino-5-methoxycarbonylbenzoic acid 96%
2-Amino-5-methoxycarbonylbenzoic acid
Formel: C₉H₉NO₄
Molecular Weight: 195,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08690064
CAS-Nummer: 63746-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR