6-Aminoquinazoline 95%

Lieferant: Apollo Scientific

OR953083-100MG OR953083-1G OR953083-250MG
APOSOR953083-100MGEA 145 EUR
APOSOR953083-100MG APOSOR953083-1G APOSOR953083-250MG
6-Aminoquinazoline 95%
6-Aminoquinazoline
Formel: C₈H₇N₃
Molecular Weight: 145,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10697883
CAS-Nummer: 101421-72-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR