2-[2-(Methylthio)phenoxy]ethylamine 95%
Lieferant: Apollo Scientific
OR953116-1G
OR953116-250MG
APOSOR953116-1GEA
632
EUR
APOSOR953116-1G
APOSOR953116-250MG
2-[2-(Methylthio)phenoxy]ethylamine 95%
2-[2-(Methylthio)phenoxy]ethylamine
Formel:
C₉H₁₃NOS Molecular Weight: 183,271 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17171366 CAS-Nummer: 72955-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...