Ethyl (s)-piperidine-2-carboxylate hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: (S)-Ethyl piperidine-2-carboxylate hydrochloride

OR953176-1G OR953176-5G
APOSOR953176-1GEA 74.1 EUR
APOSOR953176-1G APOSOR953176-5G
Ethyl (s)-piperidine-2-carboxylate hydrochloride 95%
Ethyl (s)-piperidine-2-carboxylate hydrochloride
Formel: C₈H₁₅NO₂·ClH
Molecular Weight: 193,671 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09264372
CAS-Nummer: 123495-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR