Ethyl 1-aminopyrrole-2-carboxylate hydrochloride 95%
Lieferant: Apollo Scientific
OR953189-1G
OR953189-250MG
APOSOR953189-1GEA
632
EUR
APOSOR953189-1G
APOSOR953189-250MG
Ethyl 1-aminopyrrole-2-carboxylate hydrochloride 95%
Ethyl 1-aminopyrrole-2-carboxylate hydrochloride
Formel:
C₇H₁₀N₂O₂·ClH Molecular Weight: 190.627 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12068436 CAS-Nummer: 1159825-10-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...