1-(3-Nitrophenyl)ethanol 97%

Lieferant: Apollo Scientific

OR953195-25G OR953195-5G
APOSOR953195-25GEA 237 EUR
APOSOR953195-25G APOSOR953195-5G
1-(3-Nitrophenyl)ethanol 97%
1-(3-Nitrophenyl)ethanol
Formel: C₈H₉NO₃
Molecular Weight: 167,162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00053608
CAS-Nummer: 5400-78-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR