1-(Pyridin-4-yl)propan-1-amine 95%
Lieferant: Apollo Scientific
OR953218-1G
OR953218-250MG
APOSOR953218-1GEA
208
EUR
APOSOR953218-1G
APOSOR953218-250MG
1-(Pyridin-4-yl)propan-1-amine 95%
1-(Pyridin-4-yl)propan-1-amine
Formel:
C₈H₁₂N₂ Molecular Weight: 136,194 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06637477 CAS-Nummer: 60289-68-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...