Methyl benzofuran-5-carboxylate 95%

Lieferant: Apollo Scientific

OR953290-1G OR953290-250MG
APOSOR953290-1GEA 163 EUR
APOSOR953290-1G APOSOR953290-250MG
Methyl benzofuran-5-carboxylate 95%
Methyl benzofuran-5-carboxylate
Formel: C₁₀H₈O₃
Molecular Weight: 176,169 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06204199
CAS-Nummer: 108763-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR