4-(Methylamino)cyclohexanol 97%

Lieferant: Apollo Scientific

OR953332-1G OR953332-5G
APOSOR953332-1GEA 219 EUR
APOSOR953332-1G APOSOR953332-5G
4-(Methylamino)cyclohexanol 97%
4-(Methylamino)cyclohexanol
Formel: C₇H₁₅NO
Molecular Weight: 129,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09909445
CAS-Nummer: 2987-05-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR