6-Methylpyridazine-3-carboxylic acid 95%
Lieferant: Apollo Scientific
OR953409-1G
OR953409-250MG
APOSOR953409-1GEA
348
EUR
APOSOR953409-1G
APOSOR953409-250MG
6-Methylpyridazine-3-carboxylic acid 95%
6-Methylpyridazine-3-carboxylic acid
Formel:
C₆H₆N₂O₂ Molecular Weight: 138,124 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09834366 CAS-Nummer: 64210-60-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...