6-Chloro-1,3-benzodioxol-5-amine 98%
Lieferant: Apollo Scientific
Synonyme:
6-Chlorobenzo[d][1,3]dioxol-5-amine
OR953510-1G
OR953510-250MG
APOSOR953510-1GEA
130
EUR
APOSOR953510-1G
APOSOR953510-250MG
6-Chloro-1,3-benzodioxol-5-amine 98%
6-Chloro-1,3-benzodioxol-5-amine
Formel:
C₇H₆ClNO₂ Molecular Weight: 171,581 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07643211 CAS-Nummer: 76958-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...